Learn More
Medchemexpress LLC Viloxazine hydrochloride | 35604-67-2 | MFCD00661100 | 98.9% | 273.76 | C13H20ClNO3 | 100MG
SDP
Catalog No. 5000236603
Encompass_Preferred
Viloxazine hydrochloride is a research-grade small-molecule inhibitor of the norepinephrine transporter (NET) and a modulator of serotonin (5-HT) receptors, used in preclinical studies of neurotransmitter function and attention-deficit hyperactivity disorder (ADHD). It is supplied as a white to off-white solid with molecular formula C13H20ClNO3 and a molecular weight of 273.76 g/mol. For research use only; consult the product datasheet and safety data sheet for handling, storage, and hazard information.
  • High purity (98.9%) suitable for analytical and preclinical studies.
  • White to off-white solid form for easy handling and weighing.
  • Molecular formula C13H20ClNO3 and molecular weight 273.76 g/mol.
  • Stable under recommended storage: 4°C for solid; solutions may be stored at -20°C to -80°C per guidance.
  • Datasheet and safety data sheet available for handling and regulatory compliance.
Catalog No. 50-002-36603 Supplier Medchemexpress LLC Supplier No. HY125784100MG
Viloxazine hydrochloride is a research-grade small-molecule inhibitor of the norepinephrine transporter (NET) and a modulator of serotonin (5-HT) receptors, used in preclinical studies of neurotransmitter function and attention-deficit hyperactivity disorder (ADHD). It is supplied as a white to off-white solid with molecular formula C13H20ClNO3 and a molecular weight of 273.76 g/mol. For research use only; consult the product datasheet and safety data sheet for handling, storage, and hazard information.
  • High purity (98.9%) suitable for analytical and preclinical studies.
  • White to off-white solid form for easy handling and weighing.
  • Molecular formula C13H20ClNO3 and molecular weight 273.76 g/mol.
  • Stable under recommended storage: 4°C for solid; solutions may be stored at -20°C to -80°C per guidance.
  • Datasheet and safety data sheet available for handling and regulatory compliance.