Learn More
Cayman Chemical Viloxazine (hydrochloride), Purity: ≥98%, CAS: 35604-67-2, 250 mg
SDP
Catalog No. 502914772
Encompass_Preferred
An antidepressant; selectively inhibits norepinephrine uptake over 5-HT uptake in rat hypothalamic synaptosomes (IC50s = 0.9 and >100 µM, respectively); decreases immobility time in the forced swim test in mice at 8 and 16 mg/kg
Catalog No. 50-291-4772 Supplier Cayman Chemical Supplier No. 39679250 MG
An antidepressant; selectively inhibits norepinephrine uptake over 5-HT uptake in rat hypothalamic synaptosomes (IC50s = 0.9 and >100 µM, respectively); decreases immobility time in the forced swim test in mice at 8 and 16 mg/kg