Learn More
Biocare Medical, LLC Vimentin Antibody - Concentrate, 0.1 ml
SDP
Supplier:  Biocare Medical, LLC CRM312A
Encompass_Preferred
This rabbit monoclonal Vimentin recognizes a 57-60 kDa protein, which is identified as vimentin. A study has shown that it has no crossreactivity with other closely related intermediate filament proteins such as desmin and GFAP (1). Vimentin is the main intermediate filament protein in mesenchymal cells, and therefore is of value in the differential diagnosis of undifferentiated neoplasms including melanoma and sarcoma (2).
Catalog No. 50-827-99