Learn More
enQuire Bioreagents Calponin-1 Anti-Human Monoclonal [Clone SPM169], Size: 100ug
Supplier:  enQuire Bioreagents 7431MSM3P1
Encompass_Preferred
Product Type: Antibody. Application: Flow, IF, WB, IHC. Additionally, this antibody has been validated by MS-QAVA(TM). Specificity has been quantitatively determined via Mass Spec. Conjugate/Tag: Unconjugated. Clone: VM1170. Clonality: Monoclonal. Host: Mouse. Isotype: IgG1. Positive Control: Raji, Jurkat or HeLa cells. Sarcomas or Melanomas. Cellular Staining: Cytoplasmic. Buffer: 10mM PBS with 0.05% BSA & 0.05% azide. Concentration: 200ug/ml. Purity: Protein A/G Purified. Immunogen: Recombinant full-length human vimentin protein. Storage: Short term: Store at 2-4C. Long term, aliquot and store at -20C. MW: 57-60kDa.Chromosome: 10p12.33. Gene Symbol: VIM. Gene ID: 7431. Ensemble: ENSG00000026025. Uniprot: XP_006717563.
Catalog No. 50-188-5275