Learn More
STEMCELL Technologies Anti-Human Vimentin Antibody, Clone 1A7, Size: 100 ug
SDP
Supplier:  STEMCELL Technologies 1001065
Encompass
Clone1A7 is a mouse monoclonal antibody that reacts with human vimentin a class III intermediate filament that is composed of a-helical regions linked into a coiled-coil protein and confers mechanical resistance in cells. Use the 1A7 clone in applications requiring detection of vimentin. It is verified for use with immunocytochemistry (ICC).
Catalog No. 50-239-0242