Learn More
Enzo Life Sciences Vimentin (human) monoclonal antibody (65E) (100 µl)
SDP
Supplier:  Enzo Life Sciences BMLVG61000100
Encompass_Preferred
Clone: 65E. Isotype: IgG1. Immunogen: Electrophoretically purified vimentin from human cultured fibroblasts. Species reactivity: Human. Applications: IHC, WB.
Catalog No. 50-201-2349