Learn More
Enzo Life Sciences Vimentin monoclonal antibody (1ml)
SDP
Supplier:  Enzo Life Sciences ENZABS6941000
Encompass_Preferred
Isotype: IgG. Immunogen: Recombinant vimentin. Species reactivity: Human. Applications: IHC. Purity: Affinity-purified. Formulation: Liquid. In Tris buffer, pH 7.5, containing 1% BSA and <0.1% sodium azide.
Catalog No. 50-201-3233