Learn More
Enzo Life Sciences Vimentin (pSer55) monoclonal antibody (4A4) (100µg)
SDP
Supplier:  Enzo Life Sciences ADIKAMCC249E
Encompass_Preferred
Clone: 4A4. Host: Mouse. Isotype: IgG2b. Immunogen: Synthetic peptide corresponding to a portion of mouse vimentin phosphorylated at Ser55. UniProt ID: P20152. Species reactivity: Human, Mouse, Rat. Applications: ELISA, ICC, WB. Recommended Dilutions: ELISA, Immunocytochemistry (1µg/ml) Western Blot (1-5µg/ml) Suggested dilutions/conditions may not be available for all applications. Optimal conditions must be determined individually for each application. Purity: Protein A affinity purified. Formulation: Liquid. In PBS containing 50% glycerol.
Catalog No. 50-200-7250