Learn More
Biogenex Laboratories Vimentin : V9:1mL
SDP
Supplier:  Biogenex Laboratories MU074UC
Encompass_Preferred
Vimentin is the major intermediate filament in a variety of mesenchymal or mesenchymally derived non-muscle cell types. Vimentin is found in all types of sarcomas and lymphomas. Positive staining for vimentin is seen in most cells of fibrosarcomas, liposarcomas, malignant fibrous histocytomas, angiosarcomas, chondrosarcomas and lymphomas. When the vimentin antibody is used in combination with other antibodies as a panel, it can aid in the histological classification of normal and malignant tissues.
Catalog No. 50-197-8876