Learn More
Medchemexpress LLC Vimirogant hydrochloride | 1802678-42-7 | 98.4% | 552.65 g/mol | C27H35F3N4O3S·xHCl | 1 MG
SDP
Catalog No. 5000246856
Encompass_Preferred
  • Selective RORγt inhibition: Ki = 3.5 nM and IC50 = 17 nM.
  • Reduces IL-17A secretion in multiple cell assays (reported IC50 values).
  • High purity suitable for research applications (98.39%).
  • Solid, light yellow appearance for ease of handling and storage.
  • Storage stability: keep at 4°C, sealed and away from moisture; solvent stability specified for -80°C and -20°C.
Catalog No. 50-002-46856 Supplier Medchemexpress LLC Supplier No. HY103637A1MG
  • Selective RORγt inhibition: Ki = 3.5 nM and IC50 = 17 nM.
  • Reduces IL-17A secretion in multiple cell assays (reported IC50 values).
  • High purity suitable for research applications (98.39%).
  • Solid, light yellow appearance for ease of handling and storage.
  • Storage stability: keep at 4°C, sealed and away from moisture; solvent stability specified for -80°C and -20°C.