Learn More
Medchemexpress LLC Vimseltinib | 1628606-05-2 | MFCD32858282 | 99.9% | C23H25N7O2 | 10MG
SDP
Catalog No. 5000258792
Encompass_Preferred
Vimseltinib is a small-molecule, orally bioavailable inhibitor of colony stimulating factor 1 receptor (CSF1R/c-FMS) and c-Kit intended for preclinical research. It is supplied as a characterized solid with molecular formula C23H25N7O2 and molecular weight 431.49; the certificate of analysis reports 99.94% purity (LCMS).
  • High purity for reproducible experimental results.
  • Selective inhibition of CSF1R and c-Kit kinases.
  • Suitable for in vitro and in vivo kinase studies.
  • Supplied with COA and SDS documentation for characterization.
  • Available in small research pack sizes for laboratory use.
Catalog No. 50-002-58792 Supplier Medchemexpress LLC Supplier No. HY13625610MG
Vimseltinib is a small-molecule, orally bioavailable inhibitor of colony stimulating factor 1 receptor (CSF1R/c-FMS) and c-Kit intended for preclinical research. It is supplied as a characterized solid with molecular formula C23H25N7O2 and molecular weight 431.49; the certificate of analysis reports 99.94% purity (LCMS).
  • High purity for reproducible experimental results.
  • Selective inhibition of CSF1R and c-Kit kinases.
  • Suitable for in vitro and in vivo kinase studies.
  • Supplied with COA and SDS documentation for characterization.
  • Available in small research pack sizes for laboratory use.