Learn More
Medchemexpress LLC Vimseltinib (DCC-3014) | 1628606-05-2 | 99.94% | 431.49 | 50 MG
SDP
Supplier:  Medchemexpress LLC HY13625650MG
Encompass_Preferred
Vimseltinib, also known as DCC-3014, is a c-FMS (CSF-1R) and c-Kit dual inhibitor. This compound has demonstrated anti-cancer and anti-proliferative activities with certain oral bioavailability, making it suitable for various research applications.
  • c-FMS (CSF-1R) and c-Kit dual inhibitor
  • Has anti-cancer activities
  • Has anti-proliferative activities
  • Possesses oral bioavailability
  • IC50s of <0.01 μM for c-FMS/CSF-1R
  • IC50s of 0.1-1 μM for c-Kit
Catalog No. 50-003-49150