Learn More
Biocare Medical, LLC Vina Green Chromogen Kit - 25 ml
SDP
Catalog No. NC0304477
Encompass_Preferred
Vina Green is a novel permanent chromogen that produces a green stain in the presence of horseradish peroxidase (HRP). The kit consists of liquid Vina Green chromogen and buffer that are stable, when mixed, for at least 4 hours at room temperature. Vina Green is clearly distinguishable from Warp Red and DAB on a single slide, enabling high flexibility for its application in Multiplex IHC. Vina Green is suitable for both immunohistochemistry (IHC) and in situ hybridization (ISH) applications including HPV, CMV, EBV, Kappa, Lambda, p63, HMWCK, TTF-1, Ki-67, and other targets/antigens such as blood and lymphatic vessels, and basal and myoepithelial cells. The reaction product is insoluble in alcohols and xylene and, therefore, a variety of counterstains and permanent mounting media can be used with this chromogen kit.
Catalog No. NC0304477 Supplier Biocare Medical, LLC Supplier No. BRR807AH
Vina Green is a novel permanent chromogen that produces a green stain in the presence of horseradish peroxidase (HRP). The kit consists of liquid Vina Green chromogen and buffer that are stable, when mixed, for at least 4 hours at room temperature. Vina Green is clearly distinguishable from Warp Red and DAB on a single slide, enabling high flexibility for its application in Multiplex IHC. Vina Green is suitable for both immunohistochemistry (IHC) and in situ hybridization (ISH) applications including HPV, CMV, EBV, Kappa, Lambda, p63, HMWCK, TTF-1, Ki-67, and other targets/antigens such as blood and lymphatic vessels, and basal and myoepithelial cells. The reaction product is insoluble in alcohols and xylene and, therefore, a variety of counterstains and permanent mounting media can be used with this chromogen kit.