Learn More
Medchemexpress LLC Vinaginsenoside R4 | 156009-80-2 | 1 MG
SDP
Catalog No. 5000420660
Encompass_Preferred
Vinaginsenoside R4 is isolated from the leaves of hydroponic Panax ginseng. It has an inhibitory effect on melanin biosynthesis without causing cytotoxic effects on melan-a cells, and enhances depigmentation in zebrafish.
  • Inhibits melanin biosynthesis
  • Non-cytotoxic to melan-a cells
  • Enhances depigmentation in zebrafish
Catalog No. 50-004-20660 Supplier Medchemexpress LLC Supplier No. HYN42651MG
Vinaginsenoside R4 is isolated from the leaves of hydroponic Panax ginseng. It has an inhibitory effect on melanin biosynthesis without causing cytotoxic effects on melan-a cells, and enhances depigmentation in zebrafish.
  • Inhibits melanin biosynthesis
  • Non-cytotoxic to melan-a cells
  • Enhances depigmentation in zebrafish