Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Vinaginsenoside R4 | 156009-80-2 | 5 MG
SDP

Catalog No. 5000418197
Encompass_Preferred

Vinaginsenoside R4 is a compound isolated from the leaves of hydroponic Panax ginseng. It has been shown to inhibit melanin biosynthesis without cytotoxic effects on melan-a cells and enhance depigmentation in zebrafish.

  • Purity: 99.86%
  • Molecular weight: 963.15
  • Formula: C48H82O19
  • Appearance: Solid
  • Color: White to off-white
  • Structure classification: Terpenoids, triterpenes
  • Source: Leaves of hydroponic Panax ginseng

Catalog No. 50-004-18197 Supplier Medchemexpress LLC Supplier No. HYN42655MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Vinaginsenoside R4 is a compound isolated from the leaves of hydroponic Panax ginseng. It has been shown to inhibit melanin biosynthesis without cytotoxic effects on melan-a cells and enhance depigmentation in zebrafish.

  • Purity: 99.86%
  • Molecular weight: 963.15
  • Formula: C48H82O19
  • Appearance: Solid
  • Color: White to off-white
  • Structure classification: Terpenoids, triterpenes
  • Source: Leaves of hydroponic Panax ginseng

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.