Learn More
Apexbio Technology LLC vinblastine 865-21-4 10mM (in 1mL DMSO)
SDP
Catalog No. 5000062703
Encompass_Preferred
Vinblastine is a naturally-derived alkaloid isolated from Catharanthus roseus It functions as a microtubule inhibitor by binding specifically to tubulin disrupting microtubule polymerization and thereby arresting mitosis at metaphase Due to this mechanism vinblastine is widely investigated for antitumor activities and is commonly employed in biomedical research to study cell division cytoskeletal dynamics and therapeutic interventions in oncology Vinblastine should be stored under sealed dry and cool conditions to maintain molecular integrity and biological activity
Catalog No. 50-000-62703 Supplier Apexbio Technology LLC Supplier No. N22565.1
Vinblastine is a naturally-derived alkaloid isolated from Catharanthus roseus It functions as a microtubule inhibitor by binding specifically to tubulin disrupting microtubule polymerization and thereby arresting mitosis at metaphase Due to this mechanism vinblastine is widely investigated for antitumor activities and is commonly employed in biomedical research to study cell division cytoskeletal dynamics and therapeutic interventions in oncology Vinblastine should be stored under sealed dry and cool conditions to maintain molecular integrity and biological activity