Learn More
Medchemexpress LLC HY-13780 10mg Medchemexpress, Vinblastine (sulfate) CAS:143-67-9 Purity:>98%
SDP
Catalog No. 502029031
Encompass_Preferred
Medchemexpress, HY-13780 10mg Vinblastine (sulfate) CAS:143-67-9 Vinblastine sulfate is a cytotoxic alkaloid used against various cancer types. Vinblastine sulfate inhibits the formation of microtubule and suppresses nAChR with an IC50 of 8.9 μM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-202-9031 Supplier Medchemexpress LLC Supplier No. HY1378010MG
Medchemexpress, HY-13780 10mg Vinblastine (sulfate) CAS:143-67-9 Vinblastine sulfate is a cytotoxic alkaloid used against various cancer types. Vinblastine sulfate inhibits the formation of microtubule and suppresses nAChR with an IC50 of 8.9 μM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.