Learn More
Medchemexpress LLC Vinblastine sulfate | 143-67-9 | 99.6% | 909.05 | 100 MG
SDP
Catalog No. 5000352192
Encompass_Preferred
Vinblastine sulfate is a cytotoxic alkaloid effective against various cancer types. It inhibits microtubule formation and suppresses nAChR with an IC50 of 8.9 μM. This product is intended solely for research purposes.
  • Inhibits microtubule formation and suppresses nAChR with an IC50 of 8.9 μM.
  • Blocks mitosis in HeLa cells and induces apoptosis.
  • Causes p53 alterations and DNA fragmentation, leading to apoptosis via Bax/Bcl-2 imbalance.
  • Suppresses NFκB expression and its DNA binding activity.
  • Triggers apoptosis through loss of mitochondrial membrane potential and activation of caspases.
  • Demonstrates cytotoxicity across a broad range of human cell lines.
  • Explored in various clinical trials for cancer treatment.
Catalog No. 50-003-52192 Supplier Medchemexpress LLC Supplier No. HY13780R100MG
Vinblastine sulfate is a cytotoxic alkaloid effective against various cancer types. It inhibits microtubule formation and suppresses nAChR with an IC50 of 8.9 μM. This product is intended solely for research purposes.
  • Inhibits microtubule formation and suppresses nAChR with an IC50 of 8.9 μM.
  • Blocks mitosis in HeLa cells and induces apoptosis.
  • Causes p53 alterations and DNA fragmentation, leading to apoptosis via Bax/Bcl-2 imbalance.
  • Suppresses NFκB expression and its DNA binding activity.
  • Triggers apoptosis through loss of mitochondrial membrane potential and activation of caspases.
  • Demonstrates cytotoxicity across a broad range of human cell lines.
  • Explored in various clinical trials for cancer treatment.