Learn More
Medchemexpress LLC Vinblastine sulfate | 143-67-9 | MFCD00082457 | 99.6% | C46H60N4O13S | 10MG
SDP
Catalog No. 5000258993
Encompass_Preferred
Vinblastine sulfate is an analytical reference standard of the cytotoxic alkaloid vinblastine, supplied as a solid sulfate salt for research and analytical applications. It is intended for assay development, method validation, and quality control in biochemical and pharmaceutical research, with activity as a microtubule inhibitor.
Catalog No. 50-002-58993 Supplier Medchemexpress LLC Supplier No. HY13780R10MG
Vinblastine sulfate is an analytical reference standard of the cytotoxic alkaloid vinblastine, supplied as a solid sulfate salt for research and analytical applications. It is intended for assay development, method validation, and quality control in biochemical and pharmaceutical research, with activity as a microtubule inhibitor.