Learn More
Medchemexpress LLC Vinblastine sulfate | 143-67-9 | MFCD08706468 | 99.6% | 909.05 g/mol | C46H60N4O13S | 5 MG
SDP
Catalog No. 5000222861
Encompass_Preferred
Vinblastine sulfate is an analytical standard of the cytotoxic alkaloid vinblastine, supplied as a 5 mg solid for research and analytical applications. It inhibits microtubule formation and suppresses nicotinic acetylcholine receptor activity (reported IC50 8.9 μM). The material is high purity (99.6%) and is provided with a certificate of analysis and safety data sheet for use in HPLC, GC, and MS methods.
  • High purity (99.6%) for reliable analytical results.
  • Suitable for HPLC, GC, and MS method development and validation.
  • Provided as a small solid quantity ideal for standard preparation.
  • Accompanied by certificate of analysis and safety data sheet.
  • Useful as a reference standard in quantitative and qualitative assays.
Catalog No. 50-002-22861 Supplier Medchemexpress LLC Supplier No. HY13780R5MG
Vinblastine sulfate is an analytical standard of the cytotoxic alkaloid vinblastine, supplied as a 5 mg solid for research and analytical applications. It inhibits microtubule formation and suppresses nicotinic acetylcholine receptor activity (reported IC50 8.9 μM). The material is high purity (99.6%) and is provided with a certificate of analysis and safety data sheet for use in HPLC, GC, and MS methods.
  • High purity (99.6%) for reliable analytical results.
  • Suitable for HPLC, GC, and MS method development and validation.
  • Provided as a small solid quantity ideal for standard preparation.
  • Accompanied by certificate of analysis and safety data sheet.
  • Useful as a reference standard in quantitative and qualitative assays.