Learn More
Enzo Life Sciences Vinblastine . sulfate (25mg). CAS: 143-67-9
SDP
Catalog No. 502008807
Encompass_Preferred
Plant alkaloid that arrests the cell cycle in G2/M phase by blocking mitotic spindle formation. Inhibits normal microtubule asembly and causes aberant tubulin polymerization. Blocks binding of tubulin to microtubule proteins. Induces apoptosis. Triggers Raf-1 activation, phosphorylation of Bcl-2 family proteins and induction of p53 expression. Cancer chemotherapeutic agent. Alternative name: Vincaleukoblastine . sulfate. Purity: ≥98% (HPLC). Appearance: White to light yellow crystalline powder. Solubility: Soluble in DMSO, methanol or water (25mg/ml). Long Term Storage: -20°C.
Catalog No. 50-200-8807 Supplier Enzo Life Sciences Supplier No. ALX350257M025
Plant alkaloid that arrests the cell cycle in G2/M phase by blocking mitotic spindle formation. Inhibits normal microtubule asembly and causes aberant tubulin polymerization. Blocks binding of tubulin to microtubule proteins. Induces apoptosis. Triggers Raf-1 activation, phosphorylation of Bcl-2 family proteins and induction of p53 expression. Cancer chemotherapeutic agent. Alternative name: Vincaleukoblastine . sulfate. Purity: ≥98% (HPLC). Appearance: White to light yellow crystalline powder. Solubility: Soluble in DMSO, methanol or water (25mg/ml). Long Term Storage: -20°C.