Learn More
Selleck Chemical LLC Vinblastine sulfate S4505-100mg
SDP
Catalog No. 503108326
Encompass_Preferred
Vinblastine sulfate inhibits microtubule formation and suppresses nAChR activity with IC50 of 8 9 M in a cell-free assay used to treat certain kinds of cancer Vinblastine sulfate induces autophagy and apoptosis
Catalog No. 50-310-8326 Supplier Selleck Chemical LLC Supplier No. S4505100MG
Vinblastine sulfate inhibits microtubule formation and suppresses nAChR activity with IC50 of 8 9 M in a cell-free assay used to treat certain kinds of cancer Vinblastine sulfate induces autophagy and apoptosis