Learn More
Apexbio Technology LLC Vinblastine sulfate 143-67-9 50mg
SDP
Catalog No. 5000063982
Encompass_Preferred
Vinblastine sulfate (CAS 143-67-9) is a bioactive small molecule that exerts antitumor effects primarily by disrupting microtubule formation a critical process for cell division Additionally vinblastine inhibits neuronal nicotinic acetylcholine receptor (nAChR) activity exhibiting an IC50 of 8 9 M In vitro studies demonstrated cytotoxicity with EC50 values around 15 g/mL against P815 mastocytoma cells Clinical research continues to explore vinblastine sulfate both as a monotherapy and within chemotherapeutic combinations investigating treatment protocols for metastatic melanoma and other cancers
Catalog No. 50-000-63982 Supplier Apexbio Technology LLC Supplier No. A392050
Vinblastine sulfate (CAS 143-67-9) is a bioactive small molecule that exerts antitumor effects primarily by disrupting microtubule formation a critical process for cell division Additionally vinblastine inhibits neuronal nicotinic acetylcholine receptor (nAChR) activity exhibiting an IC50 of 8 9 M In vitro studies demonstrated cytotoxicity with EC50 values around 15 g/mL against P815 mastocytoma cells Clinical research continues to explore vinblastine sulfate both as a monotherapy and within chemotherapeutic combinations investigating treatment protocols for metastatic melanoma and other cancers