Learn More
Selleck Chemical LLC Vinblastine sulfate 5mg 143-67-9 NSC49842
SDP
Supplier:  Selleck Chemical LLC S45055MG
Encompass_Preferred
Vinblastine sulfate (NSC49842) inhibits microtubule formation and suppresses nAChR activity with IC50 of 8.9 ?M in a cell-free assay, used to treat certain kinds of cancer. Vinblastine sulfate induces autophagy and apoptosis. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-136-3895