Learn More
Cayman Chemical Vinblastine (sulfate), Purity: ≥95%, CAS: 143-67-9, 50 mg
SDP
Catalog No. 502951969
Encompass_Preferred
An antimicrotubule drug used to treat Hodgkin’s lymphoma, non-small cell lung, breast, head and neck, and testicular cancer; inhibits steady-state tubulin addition to microtubules (Ki = 0.18 μM) and inhibits B16 melanoma cell proliferation (IC50 = 1 nM)
Catalog No. 50-295-1969 Supplier Cayman Chemical Supplier No. 1176250 MG
An antimicrotubule drug used to treat Hodgkin’s lymphoma, non-small cell lung, breast, head and neck, and testicular cancer; inhibits steady-state tubulin addition to microtubules (Ki = 0.18 μM) and inhibits B16 melanoma cell proliferation (IC50 = 1 nM)