Learn More
Medchemexpress LLC Vinburnine | 4880-88-0 | 99.8% | 200 MG
SDP
Catalog No. 5000414064
Encompass_Preferred
Vinburnine is a vinca alkaloid and a metabolite of vincamine, which acts as a vasodilator. It is intended for research use only.
  • Allosteric modulator of muscarinic receptors M1-M4.
  • Exhibits an anti-amnesia effect in a scopolamine-induced mouse amnesia model.
  • Promotes erythrocyte glycolysis, increases erythrocyte ATP and 2,3-diphosphoglycerol content, and improves erythrocyte deformability and tissue oxygen delivery.
Catalog No. 50-004-14064 Supplier Medchemexpress LLC Supplier No. HYB1180200MG
Vinburnine is a vinca alkaloid and a metabolite of vincamine, which acts as a vasodilator. It is intended for research use only.
  • Allosteric modulator of muscarinic receptors M1-M4.
  • Exhibits an anti-amnesia effect in a scopolamine-induced mouse amnesia model.
  • Promotes erythrocyte glycolysis, increases erythrocyte ATP and 2,3-diphosphoglycerol content, and improves erythrocyte deformability and tissue oxygen delivery.