Learn More
Cayman Chemical Vinclozolin, Purity: ≥98%, CAS: 50471-44-8, 50 mg
SDP
Catalog No. 502995320
Encompass_Preferred
A dicarboximide fungicide; active against the plant pathogenic fungi S. sclerotiorum and N. crassa (EC50s = 0.18 and 1.4 µg/ml, respectively); decreases the incidence of leaf drop in lettuce fields experimentally infected with S. sclerotiorum at 0.56 or 1.12 kg/ha
Catalog No. 50-299-5320 Supplier Cayman Chemical Supplier No. 2393950 MG
A dicarboximide fungicide; active against the plant pathogenic fungi S. sclerotiorum and N. crassa (EC50s = 0.18 and 1.4 µg/ml, respectively); decreases the incidence of leaf drop in lettuce fields experimentally infected with S. sclerotiorum at 0.56 or 1.12 kg/ha