Learn More
Selleck Chemical LLC Vincristine sulfate 50mg 2068-78-2 Leurocristine
SDP
Supplier:  Selleck Chemical LLC S124150MG
Encompass_Preferred
Vincristine sulfate (Leurocristine) is an inhibitor of polymerization of microtubules by binding to tubulin with IC50 of 32 ?M in a cell-free assay. Vincristine sulfate induces apoptosis. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-625-9