Learn More
Apexbio Technology LLC Vincristine sulfate 2068-78-2 100mg
SDP
Catalog No. 5000063095
Encompass_Preferred
Vincristine sulfate (CAS 2068-78-2) is a natural alkaloid isolated from the leaves of Catharanthus roseus structurally composed of two heterocyclic monomers vindoline and catharanthine connected by a bridging carbon It acts via inhibition of microtubule polymerization binding tubulin with a Ki of approximately 0 085 M In vitro vincristine demonstrates antiproliferative activity against B16 melanoma cells with an IC50 of 0 45 M The compound exhibits promising activity in research settings targeting diverse cancers including acute lymphoblastic leukemia acute non-lymphoblastic leukemia non-Hodgkin lymphoma Hodgkin disease and brain tumors
Catalog No. 50-000-63095 Supplier Apexbio Technology LLC Supplier No. A1765100
Vincristine sulfate (CAS 2068-78-2) is a natural alkaloid isolated from the leaves of Catharanthus roseus structurally composed of two heterocyclic monomers vindoline and catharanthine connected by a bridging carbon It acts via inhibition of microtubule polymerization binding tubulin with a Ki of approximately 0 085 M In vitro vincristine demonstrates antiproliferative activity against B16 melanoma cells with an IC50 of 0 45 M The compound exhibits promising activity in research settings targeting diverse cancers including acute lymphoblastic leukemia acute non-lymphoblastic leukemia non-Hodgkin lymphoma Hodgkin disease and brain tumors