Learn More
Medchemexpress LLC Vincristine (sulfate) | 2068-78-2 | 99.8% | 923.04 | 200 MG
SDP
Catalog No. 5000366219
Encompass_Preferred
Vincristine sulfate is an antitumor vinca alkaloid that inhibits microtubule formation in the mitotic spindle, leading to an arrest of dividing cells at the metaphase stage. It binds to microtubules with a Ki of 85 nM. This product is for research use only.
  • Antitumor vinca alkaloid
  • Inhibits microtubule formation in mitotic spindle
  • Results in arrest of dividing cells at the metaphase stage
  • Binds to microtubule with a Ki of 85 nM
  • Induces apoptosis in tumor cells
  • Inhibits SH-SY5Y cell proliferation with an IC50 of 0.1 μM
  • Induces mitotic arrest and promotes the expression of caspase-3 and -9 and cyclin B, while decreasing the expression of cyclin D
Catalog No. 50-003-66219 Supplier Medchemexpress LLC Supplier No. 200MGHYN0488
Vincristine sulfate is an antitumor vinca alkaloid that inhibits microtubule formation in the mitotic spindle, leading to an arrest of dividing cells at the metaphase stage. It binds to microtubules with a Ki of 85 nM. This product is for research use only.
  • Antitumor vinca alkaloid
  • Inhibits microtubule formation in mitotic spindle
  • Results in arrest of dividing cells at the metaphase stage
  • Binds to microtubule with a Ki of 85 nM
  • Induces apoptosis in tumor cells
  • Inhibits SH-SY5Y cell proliferation with an IC50 of 0.1 μM
  • Induces mitotic arrest and promotes the expression of caspase-3 and -9 and cyclin B, while decreasing the expression of cyclin D