Learn More
AdipoGen Vincristine sulfate
SDP
Catalog No. 502562523
Encompass_Preferred
Chemical. CAS 2068-78-2. Formula C46H56N4O10 . H2SO4. MW 825.0 . 98.1. Synthetic. Originally isolated from Vinca rosea. Immunosuppressant. Anticancer compound. Chemotherapeutic agent. Depolymerizes microtubules and irreversibly blocks binding of tubulin to microtubule proteins. Apoptosis inducer and cell cycle inhibitor in G2/M phase. Autophagy inhibitor. Inhibits microtubule-mediated fusion of autophagosome with lysosme. Inducer of IL-1beta production. Activates alone and synergistically together with doxorubicin the NLRP3-inflammasome mediated processing of IL-1beta. Monoamine oxidase MAO inhibitor.
Catalog No. 50-256-2523 Supplier AdipoGen Supplier No. AGCN20446M025
Chemical. CAS 2068-78-2. Formula C46H56N4O10 . H2SO4. MW 825.0 . 98.1. Synthetic. Originally isolated from Vinca rosea. Immunosuppressant. Anticancer compound. Chemotherapeutic agent. Depolymerizes microtubules and irreversibly blocks binding of tubulin to microtubule proteins. Apoptosis inducer and cell cycle inhibitor in G2/M phase. Autophagy inhibitor. Inhibits microtubule-mediated fusion of autophagosome with lysosme. Inducer of IL-1beta production. Activates alone and synergistically together with doxorubicin the NLRP3-inflammasome mediated processing of IL-1beta. Monoamine oxidase MAO inhibitor.