Learn More
Selleck Chemical LLC Vincristine sulfate S1241-10mM/1mL
SDP
Catalog No. 503110999
Encompass_Preferred
Vincristine sulfate is an inhibitor of polymerization of microtubules by binding to tubulin with IC50 of 32 M in a cell-free assay Vincristine sulfate induces apoptosis
Catalog No. 50-311-0999 Supplier Selleck Chemical LLC Supplier No. S124110MM/1ML
Vincristine sulfate is an inhibitor of polymerization of microtubules by binding to tubulin with IC50 of 32 M in a cell-free assay Vincristine sulfate induces apoptosis