Learn More
Medchemexpress LLC HY-N0488 50mg Medchemexpress, Vincristine (sulfate) CAS:2068-78-2 Purity:>98%
SDP
Catalog No. 502029415
Encompass_Preferred
Medchemexpress, HY-N0488 50mg Vincristine (sulfate) CAS:2068-78-2 Vincristine sulfate is an antitumor vinca alkaloid which inhibits microtubule formation in mitotic spindle, resulting in an arrest of dividing cells at the metaphase stage. It binds to microtubule with a Ki of 85 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-202-9415 Supplier Medchemexpress LLC Supplier No. HYN048850MG
Medchemexpress, HY-N0488 50mg Vincristine (sulfate) CAS:2068-78-2 Vincristine sulfate is an antitumor vinca alkaloid which inhibits microtubule formation in mitotic spindle, resulting in an arrest of dividing cells at the metaphase stage. It binds to microtubule with a Ki of 85 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.