Learn More
AdipoGen Vincristine Sulfate, 2068-78-2, MFCD08706469, 5 mg
SDP
Catalog No. 501146330
Encompass_Preferred
MF: C46H56N4O10 . H2SO4, MW: 923.044, ≥98%, Appearance: White to light-yellow powder. Immunosuppressant. Anticancer compound. Chemotherapeutic agent. Depolymerizes microtubules and irreversibly blocks binding of tubulin to microtubule proteins. Apoptosis inducer and cell cycle inhibitor in G2/M phase. Autophagy inhibitor. Inhibits microtubule-mediated fusion of autophagosome with lysosme. Inducer of IL-1beta production. Activates alone and synergistically together with doxorubicin the NLRP3-inflammasome mediated processing of IL-1beta. Monoamine oxidase (MAO) inhibitor.
Catalog No. 50-114-6330 Supplier AdipoGen Supplier No. AGCN20446M005
Add to Cart
Add to Cart
MF: C46H56N4O10 . H2SO4, MW: 923.044, ≥98%, Appearance: White to light-yellow powder. Immunosuppressant. Anticancer compound. Chemotherapeutic agent. Depolymerizes microtubules and irreversibly blocks binding of tubulin to microtubule proteins. Apoptosis inducer and cell cycle inhibitor in G2/M phase. Autophagy inhibitor. Inhibits microtubule-mediated fusion of autophagosome with lysosme. Inducer of IL-1beta production. Activates alone and synergistically together with doxorubicin the NLRP3-inflammasome mediated processing of IL-1beta. Monoamine oxidase (MAO) inhibitor.