Learn More
Cayman Chemical Vincristine (sulfate), 2068-78-2, 5 mg
SDP
Supplier:  Cayman Chemical 117645
Encompass_Preferred
Molecular Formula C46H56N4O10 • H2SO4, Purity ≥95%, Formula Weight 923.0, Synonyms: Kyocristine, Lilly 37231, Leurocristine, Novopharm, NSC 67574; An antimitotic vinca alkaloid which irreversibly blocks mitosis by binding to tubulin (Ki = 85 nM) and inhibiting tubulin polymerization; exported from cells via the ABC transporter P-glycoprotein but not by Mrp1.
Catalog No. NC0689240