Learn More
Apexbio Technology LLC Vincristine sulfate, 5mg. Cas: 2068-78-2 MFCD: MFCD08706469. Microtubule disrupter,antitumor agent
SDP
Catalog No. 501904706
Encompass_Preferred
Vincristine is a naturally occurring alkaloid that potently inhibits the tubulin addition with the inhibition constant Ki value of 0.085 M and exhibits an anti-proliferative activity against B16 melanoma cells with the half maximal inhibition concentration IC50 value of 0.45 M [1]. Other sizes are available. Please inqury us for quote.
Catalog No. 50-190-4706 Supplier Apexbio Technology LLC Supplier No. A17655
Vincristine is a naturally occurring alkaloid that potently inhibits the tubulin addition with the inhibition constant Ki value of 0.085 M and exhibits an anti-proliferative activity against B16 melanoma cells with the half maximal inhibition concentration IC50 value of 0.45 M [1]. Other sizes are available. Please inqury us for quote.