Learn More
Cayman Chemical VIncrIstInsulfate 10mg
SDP
Catalog No. 502952412
Encompass_Preferred
An antimitotic vinca alkaloid which irreversibly blocks mitosis by binding to tubulin (Ki = 85 nM) and inhibiting tubulin polymerization; exported from cells via the ABC transporter P-glycoprotein but not by Mrp1
Catalog No. 50-295-2412 Supplier Cayman Chemical Supplier No. 1176410 MG
An antimitotic vinca alkaloid which irreversibly blocks mitosis by binding to tubulin (Ki = 85 nM) and inhibiting tubulin polymerization; exported from cells via the ABC transporter P-glycoprotein but not by Mrp1