Learn More
Sino Biological Vinculin Antibody, Rabbit PAb, Antigen Affinity Purified 100uL
Supplier:  Sino Biological 201128T44100
Encompass_Preferred
Produced in rabbits immunized with E. coli-derived Human Vinculin fragment, and purified by antigen affinity chromatography.
Catalog No. 50-224-8320