Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cell Signaling Technology Vinculin Monoclonal Antibody, Rabbit IgG, Alexa Fluor™ 647, 100 μL
SDP

Catalog No. 503131862
Encompass

This rabbit monoclonal antibody is conjugated to Alexa Fluor™ 647 fluorescent dye for use in immunohistochemistry applications. Designed to recognize endogenous levels of total vinculin protein, including the 145 kDa metavinculin splice variant, it is produced using recombinant protein specific to the amino terminus of human vinculin. The conjugate is supplied at 1 mg/mL in a formulation containing phosphate buffered saline, bovine serum albumin, and less than 0.1% sodium azide.

  • Excitation and emission wavelengths of 650 nm and 668 nm, respectively
  • Validated for immunohistochemistry (paraffin) at 1:50 to 1:200 dilutions
  • Species reactivity with human, mouse, rat, and monkey samples
  • Supplied at a concentration of 1 mg/mL
  • Requires storage at 4°C protected from light; do not freeze or aliquot

Catalog No. 50-313-1862 Supplier Cell Signaling Technology Supplier No. 67324S
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This rabbit monoclonal antibody is conjugated to Alexa Fluor™ 647 fluorescent dye for use in immunohistochemistry applications. Designed to recognize endogenous levels of total vinculin protein, including the 145 kDa metavinculin splice variant, it is produced using recombinant protein specific to the amino terminus of human vinculin. The conjugate is supplied at 1 mg/mL in a formulation containing phosphate buffered saline, bovine serum albumin, and less than 0.1% sodium azide.

  • Excitation and emission wavelengths of 650 nm and 668 nm, respectively
  • Validated for immunohistochemistry (paraffin) at 1:50 to 1:200 dilutions
  • Species reactivity with human, mouse, rat, and monkey samples
  • Supplied at a concentration of 1 mg/mL
  • Requires storage at 4°C protected from light; do not freeze or aliquot

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.