Learn More
Enzo Life Sciences Vinculin (human) monoclonal antibody (FB11) (100 µl)
SDP
Supplier:  Enzo Life Sciences BMLVG61100100
Encompass_Preferred
Clone: FB11. Isotype: IgG1. Immunogen: Purified vinculin from human smooth muscle cells. Species reactivity: Human. Applications: IHC, WB. Purity: Protein A-affinity purified. Formulation: Liquid. In PBS containing 1.0% BSA and 0.09% sodium azide.
Catalog No. 50-201-2350