Learn More
TARGETMOL CHEMICALS INC Vindesine sulfate 20MG
SDP
Catalog No. 502876088
Encompass_Preferred
Also available in 1 mL, 5 mg and bulk. Please contact Fisher for quotes. Vindesine sulfate (Desacetyl Vinblastine amide) is a vinca alkaloid which is a synthetic derivative of vinblastine, binds to the microtubular proteins of the mitotic spindle, leading to crystallization of the microtubule and mitotic arrest or cell death. Purity 99.93%
Catalog No. 50-287-6088 Supplier TARGETMOL CHEMICALS INC Supplier No. T2245520MG
Also available in 1 mL, 5 mg and bulk. Please contact Fisher for quotes. Vindesine sulfate (Desacetyl Vinblastine amide) is a vinca alkaloid which is a synthetic derivative of vinblastine, binds to the microtubular proteins of the mitotic spindle, leading to crystallization of the microtubule and mitotic arrest or cell death. Purity 99.93%