Learn More
Medchemexpress LLC Vindesine sulfate | 59917-39-4 | 99.17% | 25 MG
SDP
Catalog No. 5000342358
Encompass_Preferred
Vindesine sulfate is an analytical standard intended for research and analytical applications. It functions as a potent tubulin inhibitor with a Ki of 0.110 μM, demonstrating both anti-proliferation effects in vitro and antitumor effects in vivo.
  • Analytical standard
  • Potent tubulin inhibitor
  • Anti-proliferation effects in vitro
  • Antitumor effects in vivo
  • Targets microtubule/tubulin and reference standards
  • Involved in cell cycle/DNA damage, cytoskeleton, and other pathways
Catalog No. 50-003-42358 Supplier Medchemexpress LLC Supplier No. 25MGHY129071
Vindesine sulfate is an analytical standard intended for research and analytical applications. It functions as a potent tubulin inhibitor with a Ki of 0.110 μM, demonstrating both anti-proliferation effects in vitro and antitumor effects in vivo.
  • Analytical standard
  • Potent tubulin inhibitor
  • Anti-proliferation effects in vitro
  • Antitumor effects in vivo
  • Targets microtubule/tubulin and reference standards
  • Involved in cell cycle/DNA damage, cytoskeleton, and other pathways