Learn More
Medchemexpress LLC Vindesine sulfate | 59917-39-4 | 99.4% | 50 MG
SDP
Catalog No. 5000338320
Encompass_Preferred
Vindesine sulfate is a potent tubulin inhibitor that exhibits anti-proliferation effects in vitro and antitumor effects in vivo. For example, it inhibits L-cells growth, showing approximately 25% inhibition at 40 nM.
  • Potent tubulin inhibitor with a Ki of 0.110 μM
  • Exhibits anti-proliferation effects in vitro
  • Exhibits antitumor effects in vivo
  • Inhibits L-cells growth, showing approximately 25% inhibition at 40 nM
Catalog No. 50-003-38320 Supplier Medchemexpress LLC Supplier No. HY12907150MG
Vindesine sulfate is a potent tubulin inhibitor that exhibits anti-proliferation effects in vitro and antitumor effects in vivo. For example, it inhibits L-cells growth, showing approximately 25% inhibition at 40 nM.
  • Potent tubulin inhibitor with a Ki of 0.110 μM
  • Exhibits anti-proliferation effects in vitro
  • Exhibits antitumor effects in vivo
  • Inhibits L-cells growth, showing approximately 25% inhibition at 40 nM