Learn More
Medchemexpress LLC Vindesine sulfate | 59917-39-4 | 99.40% | 10 MG
SDP
Catalog No. 5000340422
Encompass_Preferred
Vindesine sulfate is a potent tubulin inhibitor with a Ki of 0.110 μM. It demonstrates an anti-proliferation effect in vitro and an antitumor effect in vivo. In vitro, Vindesine sulfate inhibits L-cells growth, showing approximately 25% inhibition at 40 nM.
  • Potent tubulin inhibitor
  • Ki of 0.110 μM
  • Demonstrates anti-proliferation effect in vitro
  • Shows antitumor effect in vivo
  • Inhibits L-cells growth (approx. 25% inhibition at 40 nM)
Catalog No. 50-003-40422 Supplier Medchemexpress LLC Supplier No. HY12907110MG
Vindesine sulfate is a potent tubulin inhibitor with a Ki of 0.110 μM. It demonstrates an anti-proliferation effect in vitro and an antitumor effect in vivo. In vitro, Vindesine sulfate inhibits L-cells growth, showing approximately 25% inhibition at 40 nM.
  • Potent tubulin inhibitor
  • Ki of 0.110 μM
  • Demonstrates anti-proliferation effect in vitro
  • Shows antitumor effect in vivo
  • Inhibits L-cells growth (approx. 25% inhibition at 40 nM)