Learn More
Selleck Chemical LLC Vindesine sulfate S2440-25mg
SDP
Catalog No. 503112921
Encompass_Preferred
Vindesine sulfate a vinca alkaloid derived from Catharanthus roseus is a potent inhibitor of mitosis with antineoplastic activities Vindesine binds to the microtubular proteins of the mitotic spindle leading to crystallization of the microtubule and mitotic arrest or cell death
Catalog No. 50-311-2921 Supplier Selleck Chemical LLC Supplier No. S244025MG
Vindesine sulfate a vinca alkaloid derived from Catharanthus roseus is a potent inhibitor of mitosis with antineoplastic activities Vindesine binds to the microtubular proteins of the mitotic spindle leading to crystallization of the microtubule and mitotic arrest or cell death