Learn More
Enzo Life Sciences Vindoline (100mg). CAS: 2182-14-1
SDP
Supplier:  Enzo Life Sciences ALX350102M100
Encompass_Preferred
Lacks physiological activity itself, but is contained as the pentacyclic moiety in the antineoplastic agents vinblastine and vincristine. Alternative name: (2beta,3beta,4beta,5alpha,12beta,19alpha)-4-(acetyloxy)-6,7-didehydro-3-hydroxy-16-methoxy-1-methyl-Aspidospermidine-3-carboxylic acid methyl ester. Purity: ≥90% (assay on dried basis). Appearance: White to off-white crystalline powder. Solubility: Soluble in 100% ethanol and water (30mg/ml). Long Term Storage: -20°C.
Catalog No. 50-200-8733