Learn More
Apexbio Technology LLC Vindoline,20mgCAS# 2182-14-1
SDP
Supplier:  Apexbio Technology LLC N128920
Encompass_Preferred
Vindoline is a monomeric vinca-alkaloid,forms lower portion of the lead anti-tumor agents,vinblastine and vincristine. Vindoline exhibits reciprocal cross-resistance to vincristine and vinblastine. Other sizes are also available. Please inqury us for quote.
Catalog No. 50-224-0735