Learn More
Description
- Product is for educational use.
- Available in 500 mL, 1 L or 3.8 L sizes.
- Colorless Liquid
This product is for educational use - no C of A is available.
Specifications
Specifications
| Color | Colorless |
| Assay Percent Range | 5% v/v |
| Formula Weight | 60.05 |
| Concentration or Composition (by Analyte or Components) | 4 to 8% Acetic Acid Solution |
| Quantity | 3.8 L |
| Packaging | Liquid - Cider |
| Physical Form | Liquid |
| Chemical Name or Material | Vinegar |
Safety and Handling
| ShelfLife | 5 years |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
