Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC α-Viniferin | 62218-13-7 | 1 MG
SDP

Catalog No. 5000420265
Encompass_Preferred

α-Viniferin is an anti-inflammatory compound isolated from the root of *Caragana chamlagu*. It is intended for research use only. It appears as a solid with a white to yellow color, has a molecular weight of 678.68, and a chemical formula of C42H30O9.

  • Anti-inflammatory compound
  • Isolated from *Caragana chamlagu* root
  • Intended for research use only
  • Purity: 99.43%
  • Molecular weight: 678.68
  • Chemical formula: C42H30O9
  • Appearance: Solid
  • Color: White to yellow
  • Store at -20°C, protect from light
  • In solvent: Store at -80°C for 6 months; -20°C for 1 month (protect from light)
  • Classification: Phenols, polyphenols
  • Initial source: Plants other families

Catalog No. 50-004-20265 Supplier Medchemexpress LLC Supplier No. HYN28541MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

α-Viniferin is an anti-inflammatory compound isolated from the root of *Caragana chamlagu*. It is intended for research use only. It appears as a solid with a white to yellow color, has a molecular weight of 678.68, and a chemical formula of C42H30O9.

  • Anti-inflammatory compound
  • Isolated from *Caragana chamlagu* root
  • Intended for research use only
  • Purity: 99.43%
  • Molecular weight: 678.68
  • Chemical formula: C42H30O9
  • Appearance: Solid
  • Color: White to yellow
  • Store at -20°C, protect from light
  • In solvent: Store at -80°C for 6 months; -20°C for 1 month (protect from light)
  • Classification: Phenols, polyphenols
  • Initial source: Plants other families

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.