Learn More
Apexbio Technology LLC Vinorelbine 71486-22-1 10mM (in 1mL DMSO)
SDP
Supplier:  Apexbio Technology LLC N22505.1
Encompass_Preferred
Vinorelbine is a semisynthetic vinca alkaloid derived from the plant Catharanthus roseus (L ) G Don Structurally characterized by its ability to bind selectively to tubulin this compound inhibits microtubule polymerization and disrupts mitotic spindle function leading to cell cycle arrest in metaphase and subsequent apoptosis Due to these antimitotic properties vinorelbine is extensively employed in biomedical research focused on investigating cell proliferation mechanisms understanding tumor biology and evaluating cytotoxicity profiles in vitro and in vivo Commonly utilized as a chemotherapy agent it serves as a research tool in studies of cancer pharmacology and drug resistance modeling Vinorelbine must be stored in a properly sealed container under cool dry conditions to maintain its chemical stability and biological activity for experimental use
Catalog No. 50-000-63305